Methyl 4,5-difluoro-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

87146-1G 87146-500MG 87146-5G
76944-094EA 545.31 USD
76944-094 76944-096 76944-098
Methyl 4,5-difluoro-2-methylbenzoate ≥97%
Methyl 4,5-difluoro-2-methylbenzoate
Formula: C₉H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11042819
CAS Number: 1245515-60-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR