Ethyl 2-(5-cyano-2-fluorophenyl)-2-oxoacetate ≥97% research grade

Supplier: AOBChem USA

Warning

82668-1G 82668-25G 82668-500MG 82668-5G
76944-050EA 397.84 USD
76944-050 76944-052 76944-054 76944-056
Ethyl 2-(5-cyano-2-fluorophenyl)-2-oxoacetate ≥97% research grade
Ethyl 2-(5-cyano-2-fluorophenyl)-2-oxoacetate
Formula: C₁₁H₈FNO₃
Storage Temperature: Ambient
CAS Number: 1515330-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR