Ethyl 2-(4-fluoro-2-methylphenyl)-2-oxoacetate ≥97%

Supplier: AOBChem USA

Warning

82524-1G 82524-25G 82524-500MG 82524-5G
76944-034EA 131.56 USD
76944-034 76944-036 76944-038 76944-040
Ethyl 2-(4-fluoro-2-methylphenyl)-2-oxoacetate ≥97%
Ethyl 2-(4-fluoro-2-methylphenyl)-2-oxoacetate
Formula: C₁₁H₁₁FO₃
MW: 210.202 g/mol
Storage Temperature: Ambient
CAS Number: 951888-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR