Ethyl 2-(2-fluoro-5-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82501-1G 82501-25G 82501-500MG 82501-5G
76944-018EA 259.93 USD
76944-018 76944-020 76944-022 76944-024
Ethyl 2-(2-fluoro-5-(trifluoromethyl)phenyl)-2-oxoacetate ≥95%
Ethyl 2-(2-fluoro-5-(trifluoromethyl)phenyl)-2-oxoacetate
Formula: C₁₁H₈F₄O₃
MW: 264.173 g/mol
Storage Temperature: Ambient
CAS Number: 1505792-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR