Ethyl-3,4-dimethoxybenzoylformate ≥97%

Supplier: AOBChem USA

Warning

82432-1G 82432-25G 82432-500MG 82432-5G
76943-996EA 305.54 USD
76943-996 76943-998 76944-000 76944-002
Ethyl-3,4-dimethoxybenzoylformate ≥97%
Ethyl 3,4-dimethoxybenzoylformate

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₅
Storage Temperature: Ambient
CAS Number: 40233-98-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR