Ethyl 2-(4-ethoxyphenyl)-2-oxoacetate ≥95% research grade

Supplier: AOBChem USA

Warning

82427-1G 82427-25G 82427-5G
76943-982EA 83.82 USD
76943-982 76943-984 76943-986
Ethyl 2-(4-ethoxyphenyl)-2-oxoacetate ≥95% research grade
Ethyl 2-(4-ethoxyphenyl)-2-oxoacetate
Formula: C₁₂H₁₄O₄
Storage Temperature: Ambient
CAS Number: 70080-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR