Ethyl-3-methoxybenzoylformate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(3-methoxyphenyl)-2-oxoacetate

82407-25G 82407-5G
76943-972EA 456.19 USD
76943-972 76943-974
Ethyl-3-methoxybenzoylformate ≥97%
Ethyl 3-methoxybenzoylformate

Precaución: for Research Use Only

Formula: C₁₀H₉ClO₃
Storage Temperature: Ambient
CAS Number: 86358-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR