Ethyl-3-bromobenzoyl formate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-(3-bromophenyl)-2-oxoacetate

82400-1G 82400-25G 82400-500MG 82400-5G
76943-956EA 129.43 USD
76943-956 76943-958 76943-960 76943-962
Ethyl-3-bromobenzoyl formate ≥97%
Ethyl-3-bromobenzoyl formate
Formula: C₁₀H₉BrO₃
MW: 257.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07698676
CAS Number: 62123-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR