Ethyl-3-chlorobenzoylformate ≥97%

Supplier: AOBChem USA

Warning

82395-1G 82395-25G 82395-5G
76943-944EA 88.06 USD
76943-944 76943-946 76943-948
Ethyl-3-chlorobenzoylformate ≥97%
Ethyl 3-chlorobenzoylformate

Precaución: for Research Use Only

Formula: C₁₀H₉ClO₃
Storage Temperature: Ambient
MDL Number: MFCD01319616
CAS Number: 62123-73-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR