8-Fluoroquinoline-5-carboxylic acid ≥95% research grade

Supplier: AOBChem USA

Warning

80616-1G 80616-25G 80616-500MG 80616-5G
76943-930EA 334.19 USD
76943-930 76943-932 76943-934 76943-936
8-Fluoroquinoline-5-carboxylic acid ≥95% research grade
8-Fluoroquinoline-5-carboxylic acid
Formula: C₁₀H₆FNO₂
MW: 191.159 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20658731
CAS Number: 204782-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR