tert-Butyl N-(2-methylphenyl)carbamate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: tert-Butyl o-tolylcarbamate

80357-1G 80357-25G 80357-5G
76943-918EA 108.22 USD
76943-918 76943-920 76943-922
tert-Butyl N-(2-methylphenyl)carbamate ≥97%
tert-Butyl N-(2-methylphenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₂
MW: 207.269 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09031443
CAS Number: 74965-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR