N-(4-Chlorophenyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

80326-1G 80326-25G 80326-500MG 80326-5G
76943-904EA 185.66 USD
76943-904 76943-906 76943-908 76943-910
N-(4-Chlorophenyl)benzamide ≥97%
N-(4-Chlorophenyl)benzamide
Formula: C₁₃H₁₀ClNO
Storage Temperature: Ambient
CAS Number: 2866-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR