tert-Butyl 4-(5-iodopyrimidin-2-yl)piperazine-1-carboxylate ≥97%

Supplier: AOBChem USA

Warning

79855-25G 79855-5G
76943-896EA 354.35 USD
76943-896 76943-898
tert-Butyl 4-(5-iodopyrimidin-2-yl)piperazine-1-carboxylate ≥97%
tert-Butyl 4-(5-iodopyrimidin-2-yl)piperazine-1-carboxylate
Formula: C₁₃H₁₉IN₄O₂
MW: 390.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11977095
CAS Number: 1027616-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR