6-Bromo-7-methyl[1,2,4]triazolo[1,5-a]pyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Bromo-7-methyl-[1,2,4]triazolo[1,5-a]pyridine

77749-1G 77749-25G 77749-500MG 77749-5G
76943-880EA 292.81 USD
76943-880 76943-882 76943-884 76943-886
6-Bromo-7-methyl[1,2,4]triazolo[1,5-a]pyridine ≥97%
6-Bromo-7-methyl[1,2,4]triazolo[1,5-a]pyridine
Formula: C₇H₆BrN₃
MW: 212.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12026325
CAS Number: 1172534-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR