2-Bromo-3-chloro-5-hydroxypyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Bromo-5-chloropyridin-3-ol

76699-1G 76699-25G 76699-500MG 76699-5G
76943-868EA 145.35 USD
76943-868 76943-870 76943-872 76943-874
2-Bromo-3-chloro-5-hydroxypyridine ≥97%
2-Bromo-3-chloro-5-hydroxypyridine
Formula: C₅H₃BrClNO
MW: 208.4400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09835244
CAS Number: 130284-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR