Methyl 4-chloro-6-methoxynicotinate ≥97% research grade

Supplier: AOBChem USA

Warning

72920-1G 72920-25G 72920-500MG 72920-5G
76943-832EA 181.42 USD
76943-832 76943-834 76943-836 76943-838
Methyl 4-chloro-6-methoxynicotinate ≥97% research grade
Methyl 4-chloro-6-methoxynicotinate
Formula: C₈H₈ClNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD18257745
CAS Number: 848953-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR