7-Chloro-3-methyl-1H-pyrazolo[3,4-c]pyridine ≥97%

Supplier: AOBChem USA

Warning

65764-1G 65764-500MG 65764-5G
76943-790EA 792.5 USD
76943-790 76943-792 76943-794
7-Chloro-3-methyl-1H-pyrazolo[3,4-c]pyridine ≥97%
7-Chloro-3-methyl-1H-pyrazolo[3,4-c]pyridine
Formula: C₇H₆ClN₃
MW: 167.596 g/mol
Storage Temperature: Ambient
CAS Number: 2387230-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR