Ethyl isoquinoline-1-carboxylate ≥97%

Supplier: AOBChem USA

Warning

65244-25G 65244-5G
76943-780EA 559.1 USD
76943-780 76943-782
Ethyl isoquinoline-1-carboxylate ≥97%
Ethyl isoquinoline-1-carboxylate
Formula: C₁₂H₁₁NO₂
Storage Temperature: Ambient
CAS Number: 50458-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR