6-(2,2,2-Trifluoroethoxy)nicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

64950-1G 64950-25G 64950-5G
76943-766EA 79.57 USD
76943-766 76943-768 76943-770
6-(2,2,2-Trifluoroethoxy)nicotinaldehyde ≥97%
6-(2,2,2-Trifluoroethoxy)nicotinaldehyde
Formula: C8H6F3NO2
MW: 205.1300 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06738679
CAS Number: 159981-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR