6-Bromo-3-iodo-2-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

64930-1G 64930-25G 64930-500MG 64930-5G
76943-754EA 275.84 USD
76943-754 76943-756 76943-758 76943-760
6-Bromo-3-iodo-2-methoxypyridine ≥97%
6-Bromo-3-iodo-2-methoxypyridine
Formula: C₆H₅BrINO
MW: 313.9184 g/mol
Storage Temperature: Ambient
CAS Number: 1310949-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR