Methyl 3,5-dichloro-4-iodobenzoate ≥97%

Supplier: AOBChem USA

Warning

64821-1G 64821-25G 64821-500MG 64821-5G
76943-738EA 362.83 USD
76943-738 76943-740 76943-742 76943-744
Methyl 3,5-dichloro-4-iodobenzoate ≥97%
Methyl 3,5-dichloro-4-iodobenzoate
Formula: C₈H₅Cl₂IO₂
Storage Temperature: Refrigerator
CAS Number: 651058-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR