Ethyl 2,5-difluoroisonicotinate ≥97%

Supplier: AOBChem USA

Warning

64173-1G 64173-25G 64173-5G
76943-728EA 96.55 USD
76943-728 76943-730 76943-732
Ethyl 2,5-difluoroisonicotinate ≥97%
Ethyl 2,5-difluoroisonicotinate
Formula: C₈H₇F₂NO₂
Storage Temperature: Refrigerator
CAS Number: 1214387-10-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR