Ethyl 2-fluoro-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

62144-1G 62144-25G 62144-500MG 62144-5G
76943-666EA 245.07 USD
76943-666 76943-668 76943-670 76943-672
Ethyl 2-fluoro-3-methylbenzoate ≥97%
Ethyl 2-fluoro-3-methylbenzoate
Formula: C₁₀H₁₁FO₂
MW: 182.1915 g/mol
Storage Temperature: Ambient
CAS Number: 1214363-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR