6-Bromo-3-iodo-2-methylpyridine ≥95%

Supplier: AOBChem USA

Warning

61607-1G 61607-25G 61607-500MG 61607-5G
76943-636EA 165.51 USD
76943-636 76943-638 76943-640 76943-642
6-Bromo-3-iodo-2-methylpyridine ≥95%
6-Bromo-3-iodo-2-methylpyridine
Formula: C₆H₅BrIN
MW: 297.919 g/mol
Storage Temperature: Ambient
CAS Number: 1065483-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR