Methyl 2-chloro-6-fluoro-3-formylbenzoate ≥97%

Supplier: AOBChem USA

Warning

61353-1G 61353-25G 61353-500MG 61353-5G
76943-622EA 215.37 USD
76943-622 76943-624 76943-626 76943-628
Methyl 2-chloro-6-fluoro-3-formylbenzoate ≥97%
Methyl 2-chloro-6-fluoro-3-formylbenzoate
Formula: C₉H₆ClFO₃
MW: 216.5935 g/mol
Storage Temperature: Refrigerator
CAS Number: 1263283-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR