N-(6-Chloro-2-pyridinyl)-N,N-dimethylamine ≥97%

Supplier: AOBChem USA

Warning

61160-1G 61160-25G 61160-500MG 61160-5G
76943-610EA 265.23 USD
76943-610 76943-612 76943-614 76943-616
N-(6-Chloro-2-pyridinyl)-N,N-dimethylamine ≥97%
N-(6-Chloro-2-pyridinyl)-N,N-dimethylamine
Formula: C₇H₉ClN₂
Storage Temperature: Ambient
MDL Number: MFCD13188973
CAS Number: 1060801-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR