Methyl 4-formyl-2,5-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

60790-1G 60790-500MG 60790-5G
76943-586EA 593.05 USD
76943-586 76943-588 76943-590
Methyl 4-formyl-2,5-dimethylbenzoate ≥97%
Methyl 4-formyl-2,5-dimethylbenzoate
Formula: C₁₁H₁₂O₃
Storage Temperature: Refrigerator
CAS Number: 1823086-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR