6-Bromo-2-methoxy-3-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

60326-1G 60326-500MG 60326-5G
76943-566EA 657.76 USD
76943-566 76943-568 76943-570
6-Bromo-2-methoxy-3-methylpyridine ≥97%
6-Bromo-2-methoxy-3-methylpyridine
Formula: C₇H₈BrNO
Storage Temperature: Ambient
CAS Number: 1256804-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR