6-Bromo-2-fluoro-3-(trifluoromethoxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

59885-1G 59885-25G 59885-5G
76943-540EA 94.43 USD
76943-540 76943-542 76943-544
6-Bromo-2-fluoro-3-(trifluoromethoxy)benzoic acid ≥97%
6-Bromo-2-fluoro-3-(trifluoromethoxy)benzoic acid
Formula: C₈H₃BrF₄O₃
MW: 303.00 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28134151
CAS Number: 524674-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR