5-Iodo-2-methoxypyrimidine ≥97%

Supplier: AOBChem USA

Warning

59451-1G 59451-25G 59451-500MG 59451-5G
76943-524EA 134.74 USD
76943-524 76943-526 76943-528 76943-530
5-Iodo-2-methoxypyrimidine ≥97%
5-Iodo-2-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₅H₅IN₂O
Storage Temperature: Ambient
MDL Number: MFCD18412677
CAS Number: 101803-06-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR