6-Chloro-N-methylpyridine-2-carboxamide ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 6-Chloro-N-methylpicolinamide

59055-1G 59055-25G 59055-5G
76943-510EA 99.73 USD
76943-510 76943-512 76943-514
6-Chloro-N-methylpyridine-2-carboxamide ≥95%
6-Chloro-N-methylpyridine-2-carboxamide
Formula: C₇H₇ClN₂O
MW: 170.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09862945
CAS Number: 845306-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR