Potassium ethyltrifluoroborate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Potassium ethyltrifluoroboranuide

59052-1G 59052-25G 59052-5G
76943-496EA 62.6 USD
76943-496 76943-498 76943-500
Potassium ethyltrifluoroborate ≥95%
Potassium ethyltrifluoroborate
Formula: C₂H₅BF₃K
MW: 135.9656 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04112713
CAS Number: 44248-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR