Methyl 2-chloro-4-fluoro-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

58652-1G 58652-25G 58652-500MG 58652-5G
76943-486EA 393.6 USD
76943-486 76943-488 76943-490 76943-492
Methyl 2-chloro-4-fluoro-3-methylbenzoate ≥97%
Methyl 2-chloro-4-fluoro-3-methylbenzoate
Formula: C₉H₈ClFO₂
Storage Temperature: Ambient
CAS Number: 1260887-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR