Methyl 6-(p-tolyl)nicotinate ≥97%

Supplier: AOBChem USA

Warning

58045-1G 58045-25G 58045-500MG 58045-5G
76943-448EA 292.81 USD
76943-448 76943-450 76943-452 76943-454
Methyl 6-(p-tolyl)nicotinate ≥97%
Methyl 6-(p-tolyl)nicotinate
Formula: C₁₄H₁₃NO₂
MW: 227.259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28503067
CAS Number: 149467-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR