Ethyl 3-(3-bromophenyl)-3-hydroxypropanoate ≥95%

Supplier: AOBChem USA

Warning

57595-1G 57595-25G 57595-500MG 57595-5G
76943-402EA 213.25 USD
76943-402 76943-404 76943-406 76943-408
Ethyl 3-(3-bromophenyl)-3-hydroxypropanoate ≥95%
Ethyl 3-(3-bromophenyl)-3-hydroxypropanoate
Formula: C₁₁H₁₃BrO₃
Storage Temperature: Refrigerator
MDL Number: MFCD16749037
CAS Number: 51699-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR