Ethyl 4,6-difluoro-1H-indole-2-carboxylate ≥95%

Supplier: AOBChem USA

Warning

57428-1G 57428-25G 57428-500MG 57428-5G
76943-390EA 245.07 USD
76943-390 76943-392 76943-394 76943-396
Ethyl 4,6-difluoro-1H-indole-2-carboxylate ≥95%
Ethyl 4,6-difluoro-1H-indole-2-carboxylate
Formula: C₁₁H₉F₂NO₂
MW: 225.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781308
CAS Number: 870536-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR