Ethyl 2-(4-bromo-3,5-dimethyl-1H-pyrazol-1-yl)acetate ≥97% research grade

Supplier: AOBChem USA

Warning

57313-25G 57313-5G
76943-370EA 346.92 USD
76943-370 76943-372
Ethyl 2-(4-bromo-3,5-dimethyl-1H-pyrazol-1-yl)acetate ≥97% research grade
Ethyl 2-(4-bromo-3,5-dimethyl-1H-pyrazol-1-yl)acetate
Formula: C₉H₁₃BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00052541
CAS Number: 175137-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR