Methyl 1,3,5-trimethyl-1H-pyrazole-4-carboxylate ≥97%

Supplier: AOBChem USA

Warning

57241-1G 57241-25G 57241-500MG 57241-5G
76943-358EA 352.22 USD
76943-358 76943-360 76943-362 76943-364
Methyl 1,3,5-trimethyl-1H-pyrazole-4-carboxylate ≥97%
Methyl 1,3,5-trimethyl-1H-pyrazole-4-carboxylate
Formula: C₈H₁₂N₂O₂
MW: 168.193 g/mol
Storage Temperature: Ambient
CAS Number: 25016-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR