Ethyl 2-isopropylbenzoate ≥97%

Supplier: AOBChem USA

Warning

57153-1G 57153-25G 57153-500MG 57153-5G
76943-342EA 233.4 USD
76943-342 76943-344 76943-346 76943-348
Ethyl 2-isopropylbenzoate ≥97%
Ethyl 2-isopropylbenzoate
Formula: C₁₂H₁₆O₂
Storage Temperature: Ambient
CAS Number: 105337-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR