Methyl 5-acetyl-2-fluorobenzoate ≥97% research grade

Supplier: AOBChem USA

Warning

56268-1G 56268-25G 56268-500MG 56268-5G
76943-304EA 393.6 USD
76943-304 76943-306 76943-308 76943-310
Methyl 5-acetyl-2-fluorobenzoate ≥97% research grade
Methyl 5-acetyl-2-fluorobenzoate
Formula: C₁₀H₉FO₃
MW: 196.175 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26679544
CAS Number: 1031417-96-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR