Methyl 3-bromo-4-chloro-2-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

55931-1G 55931-500MG 55931-5G
76943-294EA 533.64 USD
76943-294 76943-296 76943-298
Methyl 3-bromo-4-chloro-2-fluorobenzoate ≥97%
Methyl 3-bromo-4-chloro-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅BrClFO₂
MW: 267.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28733929
CAS Number: 1804897-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR