Methyl 2-(1-hydroxycyclopentyl)acetate ≥97% research grade

Supplier: AOBChem USA

Warning

55619-1G 55619-25G 55619-500MG 55619-5G
76943-270EA 292.81 USD
76943-270 76943-272 76943-274 76943-276
Methyl 2-(1-hydroxycyclopentyl)acetate ≥97% research grade
Methyl 2-(1-hydroxycyclopentyl)acetate
Formula: C₈H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD19691252
CAS Number: 74126-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR