6-Chloro-2-fluoro-3-methoxybenzonitrile ≥95%

Supplier: AOBChem USA

Warning

55446-25G 55446-5G
76943-232EA 559.1 USD
76943-232 76943-234
6-Chloro-2-fluoro-3-methoxybenzonitrile ≥95%
6-Chloro-2-fluoro-3-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₅ClFNO
MW: 185.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832269
CAS Number: 1017777-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR