5-Iodo-2-(methylthio)pyrimidine ≥95% research grade

Supplier: AOBChem USA

Warning

54956-1G 54956-25G 54956-500MG 54956-5G
76943-192EA 245.07 USD
76943-192 76943-194 76943-196 76943-198
5-Iodo-2-(methylthio)pyrimidine ≥95% research grade
5-Iodo-2-(methylthio)pyrimidine
Formula: C₅H₅IN₂S
MW: 252.076 g/mol
Storage Temperature: Refrigerator
CAS Number: 1598164-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR