Ethyl 3-bromobenzo[b]thiophene-2-carboxylate ≥95%

Supplier: AOBChem USA

Warning

54346-1G 54346-25G 54346-500MG 54346-5G
76943-148EA 287.51 USD
76943-148 76943-150 76943-152 76943-154
Ethyl 3-bromobenzo[b]thiophene-2-carboxylate ≥95%
Ethyl 3-bromobenzo[b]thiophene-2-carboxylate
Formula: C₁₁H₉BrO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD28100541
CAS Number: 784163-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR