Methyl 4-formyl-3,5-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

53878-1G 53878-500MG 53878-5G
76943-134EA 708.69 USD
76943-134 76943-136 76943-138
Methyl 4-formyl-3,5-dimethylbenzoate ≥97%
Methyl 4-formyl-3,5-dimethylbenzoate
Formula: C₁₁H₁₂O₃
MW: 192.211 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24682163
CAS Number: 1465206-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR