Phenanthrene-9-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

53711-1G 53711-25G 53711-5G
76943-122EA 67.9 USD
76943-122 76943-124 76943-126
Phenanthrene-9-carboxylic acid ≥97%
Phenanthrene-9-carboxylic acid
Formula: C₁₅H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00019111
CAS Number: 837-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR