Methyl 2-bromo-3-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

53436-1G 53436-25G 53436-500MG 53436-5G
76943-110EA 146.41 USD
76943-110 76943-112 76943-114 76943-116
Methyl 2-bromo-3-methoxybenzoate ≥97%
Methyl 2-bromo-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
Storage Temperature: Refrigerator
MDL Number: MFCD14697991
CAS Number: 59453-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR