Methyl 5-bromo-2,3-difluoro-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

53291-1G 53291-25G 53291-500MG 53291-5G
76943-098EA 376.62 USD
76943-098 76943-100 76943-102 76943-104
Methyl 5-bromo-2,3-difluoro-4-methylbenzoate ≥97%
Methyl 5-bromo-2,3-difluoro-4-methylbenzoate
Formula: C₉H₇BrF₂O₂
MW: 265.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530207
CAS Number: 2734772-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR