Methyl 3-cyano-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

52729-1G 52729-25G 52729-500MG 52729-5G
76943-070EA 152.77 USD
76943-070 76943-072 76943-074 76943-076
Methyl 3-cyano-4-methylbenzoate ≥97%
Methyl 3-cyano-4-methylbenzoate
Formula: C₁₀H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD12405069
CAS Number: 35066-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR